Key Documents
CLS69471LBO
Pyrex® resin reaction kettle bottom
capacity 1,000 mL
Sinónimos:
pyrex kettle, reaction equipment, reaction flasks
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
wide-mouth
sterility
non-sterile
feature
closure type lid
packaging
case of 4
manufacturer/tradename
Corning 6947-1LBO
availability
not available in EU
capacity
1,000 mL
General description
Flange is finely ground for a tight seal.
Legal Information
PYREX is a registered trademark of Corning, Inc.