HPLC Analysis of Chloramphenicol in Milk on Ascentis® C18 after SPE using SupelMIP® SPE-Chloramphenicol
![HPLC Analysis of Chloramphenicol in Milk on Ascentis® C18 after SPE using SupelMIP® SPE-Chloramphenicol application for SPE, application for LC-MS](/deepweb/assets/sigmaaldrich/product/images/916/022/5d23cca5-4dd5-4dfe-9b45-995cd8f79349/800/5d23cca5-4dd5-4dfe-9b45-995cd8f79349.jpg)
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
whole pasteurized milk spiked with chloramphenicol at 15 ng/mL and 38 ng/mL
SPE tube/cartridge
SupelMIP SPE - Chloramphenicol, 25 mg/10mL (LRC) (53210-U)
condition
1 mL methanol;1 mL DI water
sample addition
1 mL
washing
2 x 1 mL MS-grade water, 1 mL 5% acetonitrile in 0.5% acetic acid, 2 x 1 mL MS-grade water, 1 mL 20% acetonitrile in 1% ammonium hydroxide. (dry SPE tubes for 15 min. under gentle vacuum; 3 x 1 mL dichloromethane; dry SPE tubes for 1 min. under gentle vacuum)
elution
elute chloramphenicol with 2 x 1 mL methanol:acetic acid:MS-grade water (89:1:10, v/v/v)
eluate post-treatment
evaporate combined eluate to dryness at 50 °C under nitrogen. reconstitute in 150 mL LC mobile phase prior to LC-MS analysis
column
Ascentis C18, 2.1 mm x 10 cm I.D., 3 μm particles (581301-U)
mobile phase
[A] 100 mM ammonium acetate: [B] MS-grade water: [C] acetonitrile (10:60:30, A:B:C)
flow rate
0.2 mL/min
column temp.
35 °C
detector
MS, ESI(-), 320-323 m/z range
injection
5 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
SupelMIP is a registered trademark of Sigma-Aldrich Co. LLC