Merck
  • Effect of ultraviolet treatment on bacterial attachment and osteogenic activity to alkali-treated titanium with nanonetwork structures.
Effect of ultraviolet treatment on bacterial attachment and osteogenic activity to alkali-treated titanium with nanonetwork structures.
International journal of nanomedicine (2017-07-20)
Honghao Zhang, Satoshi Komasa, Chiho Mashimo, Tohru Sekino, Joji Okazaki
ABSTRACT
Alkali-treated titanium with nanonetwork structures (TNS) possesses good osteogenic activity; however, the resistance of this material to bacterial contamination remains inadequate. As such, TNS implants are prone to postoperative infection. In this work, we attempted to alter the biological properties of TNS by treatment with short-duration high-intensity ultraviolet (UV) irradiation. TNS discs were treated with UV light (wavelength =254 nm, strength =100 mW/cm The superhydrophilicity and surface hydrocarbon elimination exhibited by the treated material (UV-treated titanium with a nanonetwork structure [UV-TNS]) revealed that this treatment effectively changed the surface characteristics of TNS. Notably, UV-TNS also showed reduced colonization by We suggest that simple UV treatment has great clinical potential for TNS implants, as it promotes the osseointegration of the TNS while reducing bacterial contamination, and can be conducted chair-side immediately prior to implantation.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
6-(p-Toluidino)-2-naphthalenesulfonic acid sodium salt