Merck
  • Hydrogen sulfide regulates abiotic stress tolerance and biotic stress resistance in Arabidopsis.
Hydrogen sulfide regulates abiotic stress tolerance and biotic stress resistance in Arabidopsis.
Journal of integrative plant biology (2014-10-21)
Haitao Shi, Tiantian Ye, Ning Han, Hongwu Bian, Xiaodong Liu, Zhulong Chan
ABSTRACT
Hydrogen sulfide (H2S) is an important gaseous molecule in various plant developmental processes and plant stress responses. In this study, the transgenic Arabidopsis thaliana plants with modulated expressions of two cysteine desulfhydrases, and exogenous H2S donor (sodium hydrosulfide, NaHS) and H2S scavenger (hypotaurine, HT) pre-treated plants were used to dissect the involvement of H2S in plant stress responses. The cysteine desulfhydrases overexpressing plants and NaHS pre-treated plants exhibited higher endogenous H2S level and improved abiotic stress tolerance and biotic stress resistance, while cysteine desulfhydrases knockdown plants and HT pre-treated plants displayed lower endogenous H2S level and decreased stress resistance. Moreover, H2S upregulated the transcripts of multiple abiotic and biotic stress-related genes, and inhibited reactive oxygen species (ROS) accumulation. Interestingly, MIR393-mediated auxin signaling including MIR393a/b and their target genes (TIR1, AFB1, AFB2, and AFB3) was transcriptionally regulated by H2S, and was related with H2S-induced antibacterial resistance. Moreover, H2S regulated 50 carbon metabolites including amino acids, organic acids, sugars, sugar alcohols, and aromatic amines. Taken together, these results indicated that cysteine desulfhydrase and H2S conferred abiotic stress tolerance and biotic stress resistance, via affecting the stress-related gene expressions, ROS metabolism, metabolic homeostasis, and MIR393-targeted auxin receptors.
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%