Merck
  • Development of an Australian version of the Alcohol-Related Problems Survey: a comprehensive computerised screening tool for older adults.
Development of an Australian version of the Alcohol-Related Problems Survey: a comprehensive computerised screening tool for older adults.
Australasian journal on ageing (2013-10-15)
Stephen J Bright, Arlene Fink, John C Beck, Jim Gabriel, Dhiren Singh
ABSTRACT
The Alcohol-Related Problems Survey (ARPS) reliably classifies drinking as non-hazardous, hazardous or harmful using scoring algorithms that consider quantity and frequency of alcohol use alone and in combination with health conditions, medication-use and functional status. Because it has been developed using a 14-g US standard drink, it is not valid in Australia where a standard drink contains 10 g of ethanol. We recalibrated the ARPS scoring algorithms for a 10-g Australian standard drink and updated the list of medications. The Australian ARPS (A-ARPS) was then administered to 50 non-treatment-seeking participants in waves of five. The A-ARPS recalibrated scoring algorithms reliably classified all 50 individuals. Sixty-six per cent were classified as hazardous or harmful drinkers. Many were taking medications that interact with alcohol or had medical conditions that can be exacerbated by alcohol consumption. The A-ARPS is available for use in Australia. Its utilisation could reduce the incidence of alcohol-related harms.
Sigma-Aldrich
Lisinopril, ≥98% (HPLC)
Supelco
Lisinopril, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lisinopril, United States Pharmacopeia (USP) Reference Standard