Merck
  • Effect of Pd nanoparticle size on the catalytic hydrogenation of allyl alcohol.
Effect of Pd nanoparticle size on the catalytic hydrogenation of allyl alcohol.
Journal of the American Chemical Society (2006-04-06)
Orla M Wilson, Marc R Knecht, Joaquin C Garcia-Martinez, Richard M Crooks
ABSTRACT
We report a particle size dependence for the rate of hydrogenation of allyl alcohol using 1.3-1.9 nm Pd dendrimer-encapsulated nanoparticle (DEN) catalysts. For particles with diameters of <1.5 nm and containing <147 Pd atoms, the modulation in catalytic activity is due to the electronic properties of the particle. For the larger particles, 1.5-1.9 nm in diameter and containing an average of 147-250 Pd atoms, the size effect is a result of geometrical constraints. Specifically, the hydrogenation reaction is shown to occur preferentially on the face atoms of the larger nanoparticles.
Sigma-Aldrich
Allyl alcohol, ≥99%
Sigma-Aldrich
Allyl alcohol, ≥98.5%
Sigma-Aldrich
Allyl alcohol, ≥98.5%