Merck
  • Domperidone: review of pharmacology and clinical applications in gastroenterology.
Domperidone: review of pharmacology and clinical applications in gastroenterology.
The American journal of gastroenterology (2007-05-10)
Savio C Reddymasu, Irfan Soykan, Richard W McCallum
ABSTRACT
Domperidone is a dopamine-2 receptor antagonist. It acts as an antiemetic and a prokinetic agent through its effects on the chemoreceptor trigger zone and motor function of the stomach and small intestine. Unlike metoclopramide, it does not cause any adverse neurological symptoms as it has minimal penetration through the blood-brain barrier. It thus provides an excellent safety profile for long-term administration orally in the recommended doses. Domperidone is widely used in many countries and can now be officially prescribed to patients in the United States by an investigational new drug application for the treatment of gastroparesis and any condition causing chronic nausea and vomiting. In view of this additional clinical exposure of domperidone to a new generation of gastroenterologists and other specialists, the purpose of this timely review is to revisit the pharmacology, clinical application, and safety profile of this beneficial medication.
Sigma-Aldrich
Domperidone, powder, ≥98% (HPLC)
Pricing and availability is not currently available.
Domperidone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.