Merck
  • Oxidative cleavage of alkenes using an in situ generated iodonium ion with oxone as a terminal oxidant.
Oxidative cleavage of alkenes using an in situ generated iodonium ion with oxone as a terminal oxidant.
Organic letters (2010-11-18)
Prem P Thottumkara, Thottumkara K Vinod
ABSTRACT
A facile and operationally convenient catalytic procedure for oxidative cleavage of alkenes is described. In situ formed [hydroxy(4-carboxyphenyl)iodonium]ion, 2, from the oxidation of 4-iodobenzoic acid, 1, has been shown to facilitate the cleavage of a variety of alkenes in presence of Oxone as a co-oxidant. Optimization of the reaction conditions using 1-phenyl-1-cyclohexene, 3, and the competitive oxidative cleavage of different substrates using the optimized conditions has uncovered important mechanistic details of the reaction.
Sigma-Aldrich
4-Iodobenzoic acid, 98%