Merck
  • Synthesis of N-(aminobutyl)-N-(ethylisoluminol) functionalized gold nanomaterials for chemiluminescent bio-probe.
Synthesis of N-(aminobutyl)-N-(ethylisoluminol) functionalized gold nanomaterials for chemiluminescent bio-probe.
Chemical communications (Cambridge, England) (2011-03-25)
Dayong Tian, Hongli Zhang, Ying Chai, Hua Cui
ABSTRACT
A novel strategy for the synthesis of CF-AuNMs by reducing HAuCl(4) with ABEI in aqueous solution at room temperature is reported. No additional stabilizing and reducing reagents are needed, and various morphologies of CF-AuNMs can be obtained. It could be used as bio-probe for bioassays.
Sigma-Aldrich
N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%