Merck
Structure-based discovery of inhibitors of thymidylate synthase.
Science (New York, N.Y.) (1993-03-05)
B K Shoichet, R M Stroud, D V Santi, I D Kuntz, K M Perry
ABSTRACT
A molecular docking computer program (DOCK) was used to screen the Fine Chemical Directory, a database of commercially available compounds, for molecules that are complementary to thymidylate synthase (TS), a chemotherapeutic target. Besides retrieving the substrate and several known inhibitors, DOCK proposed putative inhibitors previously unknown to bind to the enzyme. Three of these compounds inhibited Lactobacillus casei TS at submillimolar concentrations. One of these inhibitors, sulisobenzone, crystallized with TS in two configurations that differed from the DOCK-favored geometry: a counterion was bound in the substrate site, which resulted in a 6 to 9 angstrom displacement of the inhibitor. The structure of the complexes suggested another binding region in the active site that could be exploited. This region was probed with molecules sterically similar to sulisobenzone, which led to the identification of a family of phenolphthalein analogs that inhibit TS in the 1 to 30 micromolar range. These inhibitors do not resemble the substrates of the enzyme. A crystal structure of phenolphthalein with TS shows that it binds in the target site in a configuration that resembles the one suggested by DOCK.
Sigma-Aldrich
5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid, ≥97.0% (HPLC)
Supelco
Sulisobenzone, analytical standard