Merck
  • Thermo-chemical pre-treatment to solubilize and improve anaerobic biodegradability of press mud.
Thermo-chemical pre-treatment to solubilize and improve anaerobic biodegradability of press mud.
Bioresource technology (2013-01-29)
Lisbet Mailin López González, Han Vervaeren, Ileana Pereda Reyes, Ann Dumoulin, Osvaldo Romero Romero, Jo Dewulf
ABSTRACT
Different pre-treatment severities by thermo-alkaline conditions (100°C, Ca(OH)2) on press mud were evaluated for different pre-treatment time and lime loading. COD solubilization and the methane yield enhancement were assessed. The biochemical methane potential was determined in batch assays under mesophilic conditions (37±1°C). The best pre-treatment resulted in a surplus of 72% of methane yield, adding 10g Ca(OH)2 100g(-1)TS(-1) for 1h. Pre-treatment also increased the COD solubilization, but the optimal severity for COD solubilization as determined by response surface methodology did not ensure the highest methane production. Inhibitory effects on anaerobic digestion were noticed when the severity was increased. These results demonstrate the relevance of thermo-alkaline pre-treatment severity in terms of both lime loading and pre-treatment time to obtain optimal anaerobic biodegradability of lignocellulosic biomass from press mud.
Sigma-Aldrich
Calcium hydroxide, CP
Sigma-Aldrich
Calcium hydroxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium hydroxide, ACS reagent, ≥95.0%