Merck
  • Electrophilic reactivity of tetrabromorhodamine 123 is bromine induced: convergent interpretation through complementary molecular descriptors.
Electrophilic reactivity of tetrabromorhodamine 123 is bromine induced: convergent interpretation through complementary molecular descriptors.
The journal of physical chemistry. A (2012-11-09)
José A B Ferreira, Antonio Sánchez-Coronilla, Denisio M Togashi, Humberto Ferreira, José R Ascenso, Sílvia M B Costa
ABSTRACT
Nucleophilic addition of water and of methanol to 3,6-diamino-2,4,5,7-tetrabromo-9-[2-(methoxycarbonyl) phenyl]-9H-xanthen-9-ylium, 4BrR123, yields respectively 2-(3,6-diamino-2,4,5,7-tetrabromo-9-hydroxy-9H-xanthen-9-yl)xanthyl benzoate, HO4BrR123 and 2-(3,6-diamino-2,4,5,7-tetrabromo-9-methoxy-9H-xanthen-9-yl)xanthyl benzoate, MeO4BrR123. The novel experimental results are addressed theoretically. The linear free energy relationship, LFER, second-order perturbation theory analysis of the natural bond orbital, NBO, and quantum theory of atoms in molecules, QTAIM, lead to the same conclusion: the electron-withdrawing effect of bonded Br atoms in 4BrR123 extremely enhances the molecular electrophilicity, as compared to 3,6-diamino-9-[2-(methoxycarbonyl) phenyl]-9H-xanthen-9-ylium, R123. The reactivity of these diaminoxanthylium cations is discussed in the context of local and global softness in extended conjugated systems.
Sigma-Aldrich
Bromine water, CP
Sigma-Aldrich
Bromine, JIS special grade, ≥99.0%
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sigma-Aldrich
Bromine, SAJ first grade, ≥97.0%
Sigma-Aldrich
Bromine, reagent grade
Sigma-Aldrich
Bromine, ACS reagent, ≥99.5%