Merck
  • Nitrofurantoin-induced lung disease and prophylaxis of urinary tract infections.
Nitrofurantoin-induced lung disease and prophylaxis of urinary tract infections.
Primary care respiratory journal : journal of the General Practice Airways Group (2012-07-28)
Yasser Madani, Bhupinder Mann
ABSTRACT
Nitrofurantoin is commonly used for the treatment and prophylaxis of recurrent urinary tract infections (UTIs). Although relatively rare, nitrofurantoin is one of the commonest causes of drug-induced pulmonary disease, which can be potentially serious and even fatal. Knowledge of such potential adverse effects is essential to enable early recognition and withdrawal of the drug. Patients on long-term nitrofurantoin should be reviewed and monitored regularly. Management involves early consideration of the condition, and prompt withdrawal of the drug. We report three cases of nitrofurantoin-induced lung disease in patients who were on long-term nitrofurantoin for UTI prophylaxis and present a brief review of the literature on this subject.
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV)
Supelco
Nitrofurantoin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nitrofurantoin, VETRANAL®, analytical standard