Merck
  • Impact of eugenol and isoeugenol on AhR translocation, target gene expression, and proliferation in human HaCaT keratinocytes.
Impact of eugenol and isoeugenol on AhR translocation, target gene expression, and proliferation in human HaCaT keratinocytes.
Journal of toxicology and environmental health. Part A (2012-06-13)
Michaela Kalmes, Brunhilde Blömeke
ABSTRACT
The phenolic derivatives eugenol and isoeugenol, which are naturally found in essential oils of different spices, are commonly used as fragrances. Recently data demonstrated that growth suppression produced by these substances occurs in keratinocytes and that the effects may be mediated via aryl hydrocarbon receptor (AhR) interactions. In this study the effects of eugenol and isoeugenol were determined on intracellular localization of AhR, AhR target gene expression, AhR-dependent cell cycle regulation, and proliferation in HaCaT cells. Both compounds produced a rapid and marked translocation of AhR into the nucleus, induced the expression of the AhR target genes cytochrome P-450 1A1 (CYP1A1) and AhR repressor (AhRR), and inhibited proliferation of HaCaT cells. Among the G(1) phase cell cycle-related proteins, levels of the retinoblastoma protein (RB), which is known to interact with AhR, and levels of the cyclin dependent kinase (CDK) 6 were reduced by eugenol and isoeugenol, whereas steady-state levels of CDK2 and CDK4 remained unaffected. Protein levels of CDK inhibitor (CKI) p27(KIP1), known to be modulated in an AhR-dependent manner, were increased after treatment with both substances. In conclusion, data show that the antiproliferative properties of eugenol and isoeugenol in HaCaT cells are mediated through AhR, and thereby the molecular mechanisms of action in these cells were identified for the first time in this study.
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG
Supelco
Isoeugenol, analytical standard
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans