Merck
Solution-processed LiF for work function tuning in electrode bilayers.
Nano letters (2011-12-14)
Taner Aytun, Ayse Turak, Iain Baikie, Grzegorz Halek, Cleva W Ow-Yang
ABSTRACT
Although ambient processing is the key to low-cost organic solar cell production, high-vacuum thermal evaporation of LiF is often a limiting step, motivating the exploration of solution processing of LiF as an alternative electrode interlayer. Submonolayer films are realized with the assistance of polymeric micelle reactors that enable LiF particle deposition with controlled nanoscale surface coverage. Scanning Kelvin probe reveals a work function tunable with nanoparticle coverage with higher values than that of bare indium tin oxide (ITO).
Sigma-Aldrich
Lithium fluoride, powder, -300 mesh
Sigma-Aldrich
Lithium fluoride, powder, <100 μm, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium fluoride, Precipitated, 99.995%
Sigma-Aldrich
Lithium-6Li fluoride, 95 atom % 6Li, 99% (CP)