Merck
  • Assessment of nickel and cobalt release from 200 unused hand-held work tools for sale in Denmark - Sources of occupational metal contact dermatitis?
Assessment of nickel and cobalt release from 200 unused hand-held work tools for sale in Denmark - Sources of occupational metal contact dermatitis?
The Science of the total environment (2011-09-06)
Jacob P Thyssen, Peter Jensen, Carola Lidén, Anneli Julander, Morten S Jellesen, Torkil Menné, Jeanne D Johansen
ABSTRACT
Nickel and cobalt allergy remain frequent in dermatitis patients. It is important to determine possible nickel and cobalt exposures at work as these may offer important information to regulators and physicians who perform patch testing. Clinical relevance of metal exposure is usually assessed by the treating physician via the medical history and by presentation of allergic contact dermatitis. To screen unused non-powered hand-held work tools for nickel and cobalt release by using colorimetric spot tests. A random selection of 200 non-powered hand-held work tools for sale in 2 retailers of home improvement and construction products were analyzed qualitatively for metal release using the colorimetric nickel and cobalt spot tests. Nickel release was identified from 5% of 200 work tools using the dimethylglyoxime (DMG) test. In 8 of 10, positive results were located to the metal ring that acts like a cuff and is located at the end of the grip. The positive DMG test results were not related to specific categories of work tools. The cobalt spot test gave no positive test reactions. It appears that the proportion of work tools that release nickel, in amounts that may result in allergic nickel dermatitis, has decreased markedly, when results were compared with a Swedish study performed more than a decade ago. No cobalt release was detected but it should be underscored that hard-metal tools were not examined. Other sources of cobalt may explain the relatively high levels of cobalt allergy in dermatitis patients.
Sigma-Aldrich
Dimethylglyoxime, JIS special grade, ≥98.0%
Sigma-Aldrich
Dimethylglyoxime, ACS reagent, ≥99%
Sigma-Aldrich
Dimethylglyoxime, puriss. p.a., ACS reagent, for the detection of Ni, ≥99.0% (TLC)
Sigma-Aldrich
Dimethylglyoxime, ≥97.0% (TLC)