- A pterocarpan from the seeds of Bituminaria morisiana.
A pterocarpan from the seeds of Bituminaria morisiana.
Journal of natural medicines (2010-03-20)
Marco Leonti, Laura Casu, Jürg Gertsch, Leonardo Bonsignore, Costantino Floris, Mariano Casu, Filippo Cottiglia
PMID20238177
ABSTRACT
A new prenylated pterocarpan, named morisianine, was isolated together with the known secondary metabolites erybraedin C, psoralen and angelicin from the seeds of Bituminaria morisiana. The structures of the compounds were elucidated mainly by 1D and 2D NMR experiments as well as mass spectrometry. The new compound was subjected to cytotoxicity screening against a panel of human cancer cells.
MATERIALS