Merck
  • Radiochemical studies relevant to 86Y production via 86Sr(p,n)86Y for PET imaging.
Radiochemical studies relevant to 86Y production via 86Sr(p,n)86Y for PET imaging.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2008-10-22)
M Sadeghi, M Aboudzadeh, A Zali, M Mirzaii, F Bolourinovin
ABSTRACT
A novel production technique of yttrium-86 based on bombardment of deposited strontium carbonate was investigated. (86)Y was produced via proton-induced reactions on SrCO(3) target that was prepared by the sedimentation method. Production yield of 0.37mCi/microAh at 30 microA was measured by means of gamma-ray spectrometry for natural target. The separation of (86/87/88)Y from Cu and Sr was carried out by two ion-exchange columns.
Sigma-Aldrich
Strontium carbonate, ≥99.9% trace metals basis
Sigma-Aldrich
Strontium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Strontium carbonate, ≥98%