Merck
  • Maternal exposure to diethylene glycol monomethyl ether: a possible role in the etiology of retrocaval ureter.
Maternal exposure to diethylene glycol monomethyl ether: a possible role in the etiology of retrocaval ureter.
Journal of pediatric surgery (2002-08-01)
M Ihsan Karaman, Mesut Gürdal, Metin Oztürk, Hüseyin Kanberoğlu
ABSTRACT
Retrocaval ureter is a very rare condition. In light of the experimental studies, one of the etiologic factors seems to be maternal contact with diethylene glycol monomethyl ether or ethylene glycol monomethyl ether. A case of cardiovascular, skeletal, and retrocaval ureter anomalies caused by possible maternal contact while pregnant with these materials at her work in a textile factory is presented.
Sigma-Aldrich
Diethylene glycol methyl ether, SAJ first grade, ≥99.0%
Sigma-Aldrich
Diethylene glycol methyl ether, ReagentPlus®, ≥99.0%