Merck
  • Effect of dietary factors on the detection of fecal occult blood in cats.
Effect of dietary factors on the detection of fecal occult blood in cats.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc (2001-04-06)
S P Tuffli, F Gaschen, R Neiger
ABSTRACT
Eight different diets were each fed to 6 cats to evaluate the effect on a guaiac and an o-tolidine fecal occult blood test. Fecal samples were collected from day 5 through day 7. Canine blood or pure cottage cheese were used as positive and negative controls, respectively. One hundred thirty-four fecal samples were analyzed. The dry fish (capelin) and vegetable (tapioca) diet and the pure cottage cheese diet had only negative results in both tests, whereas a canned chicken and cereal (rice) diet had negative results in all fecal samples in the o-tolidine tablet test and in 10 of 16 fecal samples in the guaiac paper test. All other fecal samples from cats eating 6 other diets and the canine blood additive were positive in both fecal occult blood tests. These results indicate that occult blood tests based on o-tolidine and guaiac are clinically useful, but cats need to be on a strict diet before the tests are used.
Sigma-Aldrich
o-Tolidine solution
Sigma-Aldrich
o-Tolidine, ≥95%
Supelco
o-Tolidine, analytical standard