Merck
  • Heart rate, salivary α-amylase activity, and cooperative behavior in previously naïve children receiving dental local anesthesia.
Heart rate, salivary α-amylase activity, and cooperative behavior in previously naïve children receiving dental local anesthesia.
Pediatric dentistry (2012-12-26)
Aristidis Arhakis, George Menexes, Trilby Coolidge, Sotirios Kalfas
ABSTRACT
Psychosomatic indicators, such as heart rate (HR), salivary alpha amylase (sAA) activity, and behavior, can be used to determine stress. This study's aim was to assess the pattern of changes of salivary alpha amylase, heart rate, and cooperative behavior in previously naïve children receiving dental treatment under local anesthesia. Included were 30 children with no prior dental experience who needed 4 or more sessions of dental treatment involving local anesthesia. In each session, sAA, HR, and behavior were assessed before and during the application of local anesthesia and at the end of the treatment. The highest sAA value was always observed at the end of each session; overall, the value was lower in the fourth session. HR always increased during the local anesthesia, and did not vary across sessions. No significant relationship was found between child cooperation and either sAA or HR. In this sample, child cooperation may not be an accurate indicator of stress. Based on salivary alpha amylase activity changes, dental treatment involving local anesthesia in naïve children appeared to be less stressful after 3 sessions.
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid