Merck
  • A critical role for heme synthesis and succinate in the regulation of pluripotent states transitions.
A critical role for heme synthesis and succinate in the regulation of pluripotent states transitions.
eLife (2023-07-10)
Damien Detraux, Marino Caruso, Louise Feller, Maude Fransolet, Sébastien Meurant, Julie Mathieu, Thierry Arnould, Patricia Renard
ABSTRACT
Using embryonic stem cells (ESCs) in regenerative medicine or in disease modeling requires a complete understanding of these cells. Two main distinct developmental states of ESCs have been stabilized in vitro, a naïve pre-implantation stage and a primed post-implantation stage. Based on two recently published CRISPR-Cas9 knockout functional screens, we show here that the exit of the naïve state is impaired upon heme biosynthesis pathway blockade, linked in mESCs to the incapacity to activate MAPK- and TGFβ-dependent signaling pathways after succinate accumulation. In addition, heme synthesis inhibition promotes the acquisition of 2 cell-like cells in a heme-independent manner caused by a mitochondrial succinate accumulation and leakage out of the cell. We further demonstrate that extracellular succinate acts as a paracrine/autocrine signal, able to trigger the 2C-like reprogramming through the activation of its plasma membrane receptor, SUCNR1. Overall, this study unveils a new mechanism underlying the maintenance of pluripotency under the control of heme synthesis.
Sigma-Aldrich
Monoclonal Anti-5-Methylcytosine / 5-mC antibody produced in mouse, clone GT4111, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Zscan4 Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.