Merck
  • Peptidase inhibitor (PI16) impairs bladder cancer metastasis by inhibiting NF-κB activation via disrupting multiple-site ubiquitination of NEMO.
Peptidase inhibitor (PI16) impairs bladder cancer metastasis by inhibiting NF-κB activation via disrupting multiple-site ubiquitination of NEMO.
Cellular & molecular biology letters (2023-08-01)
Xiangqin Kuang, Zhuojun Zhang, Difeng Li, Wenhao Bao, Jinyuan Pan, Ping Zhou, Han Chen, Zhiqing Gao, Xiaoyi Xie, Chunxiao Yang, Ge Zhu, Zhongqiu Zhou, Ruiming Tang, Zhengfu Feng, Lihuan Zhou, Xiaoli Feng, Lan Wang, Jianan Yang, Lili Jiang
ABSTRACT
Bladder cancer (BLCA) is a malignancy that frequently metastasizes and leads to poor patient prognosis. It is essential to understand the molecular mechanisms underlying the progression and metastasis of BLCA and identify potential biomarkers. The expression of peptidase inhibitor 16 (PI16) was analysed using quantitative PCR, immunoblotting and immunohistochemistry assays. The functional roles of PI16 were evaluated using wound healing, transwell, and human umbilical vein endothelial cell tube formation assays, as well as in vivo tumour models. The effects of PI16 on nuclear factor κB (NF-κB) signalling activation were examined using luciferase reporter gene systems, immunoblotting and immunofluorescence assays. Co-immunoprecipitation was used to investigate the interaction of PI16 with annexin-A1 (ANXA1) and NEMO. PI16 expression was downregulated in bladder cancer tissues, and lower PI16 levels correlated with disease progression and poor survival in patients with BLCA. Overexpressing PI16 inhibited BLCA cell growth, motility, invasion and angiogenesis in vitro and in vivo, while silencing PI16 had the opposite effects. Mechanistically, PI16 inhibited the activation of the NF-κB pathway by interacting with ANXA1, which inhibited K63 and M1 ubiquitination of NEMO. These results indicate that PI16 functions as a tumour suppressor in BLCA by inhibiting tumour growth and metastasis. Additionally, PI16 may serve as a potential biomarker for metastatic BLCA.
Sigma-Aldrich
Anti-Linear Ubiquitin Antibody, clone LUB9, clone LUB9, from mouse
Pricing and availability is not currently available.