Merck
  • Cervical Cancer Cells-Derived Extracellular Vesicles Containing microRNA-146a-5p Affect Actin Dynamics to Promote Cervical Cancer Metastasis by Activating the Hippo-YAP Signaling Pathway via WWC2.
Cervical Cancer Cells-Derived Extracellular Vesicles Containing microRNA-146a-5p Affect Actin Dynamics to Promote Cervical Cancer Metastasis by Activating the Hippo-YAP Signaling Pathway via WWC2.
Journal of oncology (2022-07-09)
Weiwei Wang, Lipei Wu, Jiale Tian, Wenhui Yan, Chunrun Qi, Wuchao Liu, Shihai Xuan, Anquan Shang
ABSTRACT
Application of extracellular vesicles (EVs) for cancer treatment has been well-documented. We probed into the potential role of cervical cancer cells-secreted EVs by transferring miR-146a-5p in cervical cancer. After characterization of miR-146a-5p expression in clinical cervical cancer tissue samples, gain- and loss-of-function experiments were implemented to test the effect of miR-146a-5p on the invasion, epithelial-mesenchymal transition (EMT), and anoikis in cervical cancer cells. EVs were isolated from high-metastatic cervical cancer cells, after which their effects on the malignant behaviors of low-metastatic cervical cancer cells were assessed in a co-culture system. Luciferase assay was implemented to validate the putative binding relationship between miR-146a-5p and WWC2, followed by further investigation of downstream pathway (Hippo-YAP). Finally, nude mouse lung metastasis model was developed for in vivo validation. miR-146a-5p was elevated in cervical cancer tissues and high miR-146a-5p expression promoted the metastatic potential of cervical cancer cells through enhancing their invasiveness and anoikis resistance, and inducing EMT. Furthermore, miR-146a-5p carried by EVs secreted by highly metastatic cervical cancer cells could promote the metastasis of low-metastatic cervical cancer cells. Mechanistically, miR-146a-5p targeted WWC2 to activate YAP, by which it inhibited the phosphorylation of cofilin, and promoted the process of cofilin-mediated depolymerization of F-actin to G-actin. In vivo data demonstrated that EVs-carried miR-146a-5p promoted tumor metastasis through the WWC2/YAP axis. Cancer-derived EVs delivered pro-metastatic miR-146a-5p to regulate the actin dynamics in cervical cancer, thereby leading to cancer metastasis. This experiment highlighted an appealing therapeutic modality for cervical cancer.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Pricing and availability is not currently available.
Roche
Fibronectin, from human plasma
Pricing and availability is not currently available.