Merck
  • Survivin and XIAP - two potential biological targets in follicular thyroid carcinoma.
Survivin and XIAP - two potential biological targets in follicular thyroid carcinoma.
Scientific reports (2017-09-14)
Thomas A Werner, Levent Dizdar, Inga Nolten, Jasmin C Riemer, Sabrina Mersch, Sina C Schütte, Christiane Driemel, Pablo E Verde, Katharina Raba, Stefan A Topp, Matthias Schott, Wolfram T Knoefel, Andreas Krieg
ABSTRACT
Follicular thyroid carcinoma's (FTC) overall good prognosis deteriorates if the tumour fails to retain radioactive iodine. Therefore, new druggable targets are in high demand for this subset of patients. Here, we investigated the prognostic and biological role of survivin and XIAP in FTC. Survivin and XIAP expression was investigated in 44 FTC and corresponding non-neoplastic thyroid specimens using tissue microarrays. Inhibition of both inhibitor of apoptosis proteins (IAP) was induced by shRNAs or specific small molecule antagonists and functional changes were investigated in vitro and in vivo. Survivin and XIAP were solely expressed in FTC tissue. Survivin expression correlated with an advanced tumour stage and recurrent disease. In addition, survivin proved to be an independent negative prognostic marker. Survivin or XIAP knockdown caused a significant reduction in cell viability and proliferation, activated caspase3/7 and was associated with a reduced tumour growth in vivo. IAP-targeting compounds induced a decrease of cell viability, proliferation and cell cycle activity accompanied by an increase in apoptosis. Additionally, YM155 a small molecule inhibitor of survivin expression significantly inhibited tumour growth in vivo. Both IAPs demonstrate significant functional implications in the oncogenesis of FTCs and thus prove to be viable targets in patients with advanced FTC.
Product Description
FTC-133 Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.