Merck
  • Experimental study of fat derived pellets promoting wound healing in rats.
Experimental study of fat derived pellets promoting wound healing in rats.
Bioengineered (2021-11-18)
Li Zeng, Shaolong Zhou, Chen Chen, Lin-Hua Zhou, Xiujiang Shi, Zongjian Wu, Sheng-Kang Luo
ABSTRACT
To observe the effect of fat-derived pellets (FDP) on wound healing in rats, the inguinal fat of rats was obtained, and the FDP were obtained after centrifugation. The cell activity and growth factor secretion of FDP were measured. The wounds in rats were created, and FDP was used to treat the wounds of rats. The phenotype of macrophages and the expression of angiogenic factors expression in wounds were measured. The cell viability in FDP remains in high level after centrifugation and the expression of vascular endothelial growth factor (VEGF) and Basic Fibroblast Growth Factor (bFGF) from FDP was observed in vitro. The FDP significantly promoted the wound healing of rats compared with that in control groups. Moreover, the expression of M2 macrophages and VEGF in FDP group were significantly higher than that in the control group. FDP is a kind of stem cell product, which can be obtained from adipose tissue by physical centrifugation. The cytotherapeutic effect of FDP makes it a promising product for wound healing in clinics.
Sigma-Aldrich
Rat VEGF ELISA Kit, for serum, plasma and cell culture supernatant
Pricing and availability is not currently available.
Sigma-Aldrich
Mouse bFGF ELISA Kit, for serum, plasma and cell culture supernatant
Pricing and availability is not currently available.