Merck
  • T and B cell abnormalities, pneumocystis pneumonia, and chronic lymphocytic leukemia associated with an AIOLOS defect in patients.
T and B cell abnormalities, pneumocystis pneumonia, and chronic lymphocytic leukemia associated with an AIOLOS defect in patients.
The Journal of experimental medicine (2021-10-26)
Hye Sun Kuehn, Jingjie Chang, Motoi Yamashita, Julie E Niemela, Chengcheng Zou, Kazuki Okuyama, Junji Harada, Jennifer L Stoddard, Cristiane J Nunes-Santos, Brigette Boast, Ryan M Baxter, Elena W Y Hsieh, Mary Garofalo, Thomas A Fleisher, Tomohiro Morio, Ichiro Taniuchi, Cullen M Dutmer, Sergio D Rosenzweig
ABSTRACT
AIOLOS/IKZF3 is a member of the IKAROS family of transcription factors. IKAROS/IKZF1 mutations have been previously associated with different forms of primary immunodeficiency. Here we describe a novel combined immunodeficiency due to an IKZF3 mutation in a family presenting with T and B cell involvement, Pneumocystis jirovecii pneumonia, and/or chronic lymphocytic leukemia. Patients carrying the AIOLOS p.N160S heterozygous variant displayed impaired humoral responses, abnormal B cell development (high percentage of CD21low B cells and negative CD23 expression), and abrogated CD40 responses. Naive T cells were increased, T cell differentiation was abnormal, and CD40L expression was dysregulated. In vitro studies demonstrated that the mutant protein failed DNA binding and pericentromeric targeting. The mutant was fully penetrant and had a dominant-negative effect over WT AIOLOS but not WT IKAROS. The human immunophenotype was recapitulated in a murine model carrying the corresponding human mutation. As demonstrated here, AIOLOS plays a key role in T and B cell development in humans, and the particular gene variant described is strongly associated with immunodeficiency and likely malignancy.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Anti-Aiolos Antibody, clone 9D10, clone 9D10, from mouse
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder