Merck
  • Synthesis and Characterization of New Zinc Phthalocyanine - Dodecenyl Succinic Anhydride Benzoic Groups.
Synthesis and Characterization of New Zinc Phthalocyanine - Dodecenyl Succinic Anhydride Benzoic Groups.
Current organic synthesis (2020-05-20)
Hussein Ali Al-Bahrani, Mohanad Mousa Kareem, Abdul Amir Kadhum, Nour A Alrazzak
ABSTRACT
The phthalocyanines a series of compounds involves four iso-indole units linked by aza nitrogen atoms bonded with metal atoms that are normally located in the center a phthalocyanines ring. Some of the central metal-phthalocyanines can be excited by ultraviolet light and emit a fluorescence in far-red region. To synthesize a derivative of phthalocyanines namely 4,4',4' '-tri-(dodecenyl succinic anhydride)- 4' ' '-(5-amino salicylic acid) zinc phthalocyanine with a zinc central metal. The reaction of 4- nitro Phthalonitrile and 4- amino Phthalonitrile with ZnCl2 in the presence of dimethyl amino ethanol afforded 4,4',4' '-triamino-4' ' '-nitro zinc phthalocyanine. This product reacted with 5-amino salicylic acid to yield tetra-(5-amino salicylic acid) zinc phthalocyanine. A dodecenyl succinic anhydride was added on the amine group of benzoic rings to afford 4,4',4' '-tri-(dodecenyl succinic anhydride)-4' ' '-(5-amino salicylic acid) zinc phthalocyanine(I), the target compound. Compound I is successfully synthesized with a yield of 72% from tetra-(5-amino salicylic acid) zinc phthalocyanine with dodecenyl succinic anhydride. The newly synthesized molecule of 4,4',4' '-tri-(dodecenyl succinic anhydride)-4' ' '-(5-amino salicylic acid) zinc phthalocyanine (I), tetra-(5-amino salicylic acid) zinc phthalocyanine(E) and 4,4',4' '- triamino-4' ' '-nitro zinc phthalocyanine (S). The reaction of 4- nitro Phthalonitrile and 4- amino and the structure of compound I is confirmed and its formation was proven.
Sigma-Aldrich
4-Nitrophthalonitrile, 99%