Merck
  • Divergent Roles for Macrophage C-type Lectin Receptors, Dectin-1 and Mannose Receptors, in the Intestinal Inflammatory Response.
Divergent Roles for Macrophage C-type Lectin Receptors, Dectin-1 and Mannose Receptors, in the Intestinal Inflammatory Response.
Cell reports (2020-04-03)
Mouna Rahabi, Godefroy Jacquemin, Mélissa Prat, Etienne Meunier, Mohamad AlaEddine, Bénédicte Bertrand, Lise Lefèvre, Khaddouj Benmoussa, Philippe Batigne, Agnès Aubouy, Johan Auwerx, Sylvain Kirzin, Delphine Bonnet, Marie Danjoux, Bernard Pipy, Laurent Alric, Hélène Authier, Agnès Coste
ABSTRACT
Colonic macrophages are considered to be major effectors of inflammatory bowel diseases (IBDs) and the control of gut inflammation through C-type lectin receptors is an emerging concept. We show that during colitis, the loss of dectin-1 on myeloid cells prevents intestinal inflammation, while the lack of mannose receptor (MR) exacerbates it. A marked increase in dectin-1 expression in dextran sulfate sodium (DSS)-exposed MR-deficient mice supports the critical contribution of dectin-1 to colitis outcome. Dectin-1 is crucial for Ly6ChighCCR2high monocyte population enrichment in the blood and their recruitment to inflamed colon as precursors of inflammatory macrophages. Dectin-1 also promotes inflammasome-dependent interleukin-1β (IL-1β) secretion through leukotriene B4 production. Interestingly, colonic inflammation is associated with a concomitant overexpression of dectin-1/CCL2/LTA4H and downregulation of MR on macrophages from IBD patients. Thus, MR and dectin-1 on macrophages are important mucosal inflammatory regulators that contribute to the intestinal inflammation.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Zymosan A from Saccharomyces cerevisiae, for inducing inflamatory response
Roche
High Pure PCR Template Preparation Kit, pkg of 100 purifications, suitable for DNA extraction
Supelco
Aprobarbital solution, 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
Caspase-1 Inhibitor VI, Caspase-1 Inhibitor VI, Z-YVAD-FMK, is a potent, cell-permeable, and irreversible inhibitor of caspase-1, caspase-4, and caspase-5.