Merck
  • Modulation of T lymphocyte replicative senescence via TNF-{alpha} inhibition: role of caspase-3.
Modulation of T lymphocyte replicative senescence via TNF-{alpha} inhibition: role of caspase-3.
Journal of immunology (Baltimore, Md. : 1950) (2009-03-21)
Stanley T Parish, Jennifer E Wu, Rita B Effros
ABSTRACT
Expanded populations of CD8(+) T lymphocytes lacking CD28 expression are associated with a variety of deleterious clinical outcomes, including early mortality in the elderly, more rapid progression to AIDS, cardiovascular disease, and enhanced tumor cell growth. In cell culture, irreversible loss of CD28 expression correlates with increased production of TNF-alpha as CD8(+) T cells are driven to the nonproliferative end stage of replicative senescence by multiple rounds of Ag-driven cell division. Interestingly, in patients with rheumatoid arthritis, inhibition or neutralization of TNF-alpha reduces the proportion of T cells lacking CD28 in the disease joints, consistent with studies showing a direct involvement of this cytokine in CD28 gene transcription. Here, we show that modulation of TNF-alpha levels in long-term cultures of human CD8(+) T lymphocytes, by chronic exposure either to a neutralizing Ab or to an inhibitor of the TNF-alpha receptor-1, increases proliferative potential, delays loss of CD28 expression, retards cytokine profile changes, and enhances telomerase activity. We also show that constitutive caspase-3, one of the downstream effectors of TNF-alphaR1 binding, increases in parallel with the loss of CD28 in long-term cultures, but this effect is blunted in the presence of the TNF-alpha inhibitors. Consistent with the in vitro culture data, CD8(+)CD28(-) T lymphocytes tested immediately ex vivo also show significantly higher levels of caspase-3 compared with their CD28(+) counterparts. These findings help elucidate the complex nature of CD28 gene regulation, and may ultimately lead to novel therapeutic approaches for diseases associated with increased proportions of CD28(-) T lymphocytes.
Sigma-Aldrich
TRAPeze RT Telomerase Detection Kit, A highly sensitive in vitro assay for the fluorometric detection & real time quantification of telomerase activity in cells.
Pricing and availability is not currently available.