Merck
  • Activation of NF-kappaB signaling by inhibitor of NF-kappaB kinase beta increases aggressiveness of ovarian cancer.
Activation of NF-kappaB signaling by inhibitor of NF-kappaB kinase beta increases aggressiveness of ovarian cancer.
Cancer research (2010-04-29)
Lídia Hernandez, Sarah C Hsu, Ben Davidson, Michael J Birrer, Elise C Kohn, Christina M Annunziata
ABSTRACT
The NF-kappaB family of transcription factors has been implicated in the propagation of ovarian cancer, but the significance of constitutive NF-kappaB signaling in ovarian cancer is unknown. We hypothesized that constitutive NF-kappaB signaling defines a subset of ovarian cancer susceptible to therapeutic targeting of this pathway. We investigated the biological relevance of NF-kappaB in ovarian cancer using a small-molecule inhibitor of inhibitor of NF-kappaB kinase beta (IKKbeta) and confirmed with RNA interference toward IKKbeta. We developed a gene expression signature of IKKbeta signaling in ovarian cancer using both pharmacologic and genetic manipulation of IKKbeta. The expression of IKKbeta protein itself and the nine-gene ovarian cancer-specific IKKbeta signature were related to poor outcome in independently collected sets of primary ovarian cancers (P = 0.02). IKKbeta signaling in ovarian cancer regulated the transcription of genes involved in a wide range of cellular effects known to increase the aggressive nature of the cells. We functionally validated the effect of IKKbeta signaling on proliferation, invasion, and adhesion. Downregulating IKKbeta activity, either by a small-molecule kinase inhibitor or by short hairpin RNA depletion of IKKbeta, blocked all of these cellular functions, reflecting the negative regulation of the target genes identified. The diversity of functions controlled by IKKbeta in ovarian cancer suggests that therapeutic blockade of this pathway could be efficacious if specific IKKbeta inhibitor therapy is focused to patients whose tumors express a molecular profile suggestive of dependence on IKKbeta activity.
Sigma-Aldrich
IKK-2 Inhibitor IV, The IKK-2 Inhibitor IV, also referenced under CAS 507475-17-4, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-IKKε Antibody, Upstate®, from rabbit