Merck
  • Hsp90 regulates HDAC3-dependent gene transcription while HDAC3 regulates the functions of Hsp90.
Hsp90 regulates HDAC3-dependent gene transcription while HDAC3 regulates the functions of Hsp90.
Cellular signalling (2020-10-06)
Akhil Kotwal, Sreedhar Amere Subbarao
ABSTRACT
Deregulated DNA methylation and post-translational histone modifications are majorly associated with cancer progression. Histone modification regulates the gene expression patterns that contribute to the emergence of sporadic cancers. Histone deacetylases (HDACs) act as erasers of acetylation marks, and their functions are often deregulated in cancer. Since non-histone proteins can also act as substrates for HDACs, identifying their involvement in vital regulatory molecules contributing to cancer progression is essential. Hsp90 is a cancer chaperone that contributes to kinase evolution and, thus, cellular adaptations. Acetylated Hsp90 loses its chaperoning functions and client protein interactions. Robust cell proliferation is one of the hallmarks of cancer. However, Hsp90 involvement in cancer promoting gene transcription is less understood. Using human breast cancer cells, we demonstrate that nuclear Hsp90 functions are regulated by HDAC3, while Hsp90 regulates HDAC3 nuclear translocation. Pharmacological inhibition of Hsp90 decreased the HDAC3 nuclear translocation and increased the gene expression relevant to epithelial to mesenchymal transition. Further, inhibition of HDAC3 resulted in the nuclear accumulation of acetylated Hsp90. Additionally, Hsp90 inhibition affected the global histone acetylation and methylation patterns, whereas HDAC3 inhibition exhibited less impact. Our results display a novel regulatory mechanism mediated by Hsp90 and HDAC3 in tumor cells. Considering that Hsp90 and histone deacetylase inhibitors are emerging as novel anticancer agents, our findings may have clinical relevance.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Hamilton® syringe, luer tip, 705LT, volume 50 μL, needle size (not included)