Merck
Profile of Back Bacon Produced From the Common Warthog.
Foods (Basel, Switzerland) (2020-05-21)
Louwrens C Hoffman, Monlee Rudman, Alison J Leslie
ABSTRACT
The common warthog (Phacochoerus africanus) has historically been hunted and consumed by rural communities throughout its distribution range in Africa. This study aims to develop a processed product from warthog meat in the form of back bacon (Longissimus thoracis et lumborum) as a healthy alternative meat product and to determine its chemical and sensory characteristics derived from adult and juvenile boars and sows. The highest scored attributes included typical bacon and smoky aroma and flavor, and salty flavor, as well as tenderness and juiciness. Neither sex nor age influenced the bacon's chemical composition; the bacon was high in protein (~29%) and low in total fat (<2%). Palmitic (C16:0), stearic (C18:0), linoleic (C18:2ω6), oleic (C18:1ω9c), and arachidonic (C20:4ω6) were the dominant fatty acids. There was an interaction between sex and age for the PUFA:SFA ratio (p = 0.01). The differences between sex and age class are considered negligible regarding the overall profile and healthiness of the bacon.
Sigma-Aldrich
Heptadecanoic acid, ≥98%
Pricing and availability is not currently available.