Merck
  • Rational Design of Bioavailable Photosensitizers for Manipulation and Imaging of Biological Systems.
Rational Design of Bioavailable Photosensitizers for Manipulation and Imaging of Biological Systems.
Cell chemical biology (2020-07-23)
Thomas C Binns, Anthony X Ayala, Jonathan B Grimm, Ariana N Tkachuk, Guillaume A Castillon, Sebastien Phan, Lixia Zhang, Timothy A Brown, Zhe Liu, Stephen R Adams, Mark H Ellisman, Minoru Koyama, Luke D Lavis
ABSTRACT
Light-mediated chemical reactions are powerful methods for manipulating and interrogating biological systems. Photosensitizers, compounds that generate reactive oxygen species upon excitation with light, can be utilized for numerous biological experiments, but the repertoire of bioavailable photosensitizers is limited. Here, we describe the synthesis, characterization, and utility of two photosensitizers based upon the widely used rhodamine scaffold and demonstrate their efficacy for chromophore-assisted light inactivation, cell ablation in culture and in vivo, and photopolymerization of diaminobenzidine for electron microscopy. These chemical tools will facilitate a broad range of applications spanning from targeted destruction of proteins to high-resolution imaging.
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Durcupan ACM, set for 1 L embedding mixture