Merck
  • Antitumor activity of a systemic STING-activating non-nucleotide cGAMP mimetic.
Antitumor activity of a systemic STING-activating non-nucleotide cGAMP mimetic.
Science (New York, N.Y.) (2020-08-21)
Emily N Chin, Chenguang Yu, Vincent F Vartabedian, Ying Jia, Manoj Kumar, Ana M Gamo, William Vernier, Sabrina H Ali, Mildred Kissai, Daniel C Lazar, Nhan Nguyen, Laura E Pereira, Brent Benish, Ashley K Woods, Sean B Joseph, Alan Chu, Kristen A Johnson, Philipp N Sander, Francisco Martínez-Peña, Eric N Hampton, Travis S Young, Dennis W Wolan, Arnab K Chatterjee, Peter G Schultz, H Michael Petrassi, John R Teijaro, Luke L Lairson
ABSTRACT
Stimulator of interferon genes (STING) links innate immunity to biological processes ranging from antitumor immunity to microbiome homeostasis. Mechanistic understanding of the anticancer potential for STING receptor activation is currently limited by metabolic instability of the natural cyclic dinucleotide (CDN) ligands. From a pathway-targeted cell-based screen, we identified a non-nucleotide, small-molecule STING agonist, termed SR-717, that demonstrates broad interspecies and interallelic specificity. A 1.8-angstrom cocrystal structure revealed that SR-717 functions as a direct cyclic guanosine monophosphate-adenosine monophosphate (cGAMP) mimetic that induces the same "closed" conformation of STING. SR-717 displayed antitumor activity; promoted the activation of CD8+ T, natural killer, and dendritic cells in relevant tissues; and facilitated antigen cross-priming. SR-717 also induced the expression of clinically relevant targets, including programmed cell death 1 ligand 1 (PD-L1), in a STING-dependent manner.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
SR-301, ≥98% (HPLC)
Sigma-Aldrich
Cyclic-di-GMP sodium salt, ≥98% (HPLC)