Merck
  • Analysis of α-synuclein species enriched from cerebral cortex of humans with sporadic dementia with Lewy bodies.
Analysis of α-synuclein species enriched from cerebral cortex of humans with sporadic dementia with Lewy bodies.
Brain communications (2020-04-14)
John B Sanderson, Suman De, Haiyang Jiang, Matteo Rovere, Ming Jin, Ludovica Zaccagnini, Aurelia Hays Watson, Laura De Boni, Valentina N Lagomarsino, Tracy L Young-Pearse, Xinyue Liu, Thomas C Pochapsky, Bradley T Hyman, Dennis W Dickson, David Klenerman, Dennis J Selkoe, Tim Bartels
ABSTRACT
Since researchers identified α-synuclein as the principal component of Lewy bodies and Lewy neurites, studies have suggested that it plays a causative role in the pathogenesis of dementia with Lewy bodies and other 'synucleinopathies'. While α-synuclein dyshomeostasis likely contributes to the neurodegeneration associated with the synucleinopathies, few direct biochemical analyses of α-synuclein from diseased human brain tissue currently exist. In this study, we analysed sequential protein extracts from a substantial number of patients with neuropathological diagnoses of dementia with Lewy bodies and corresponding controls, detecting a shift of cytosolic and membrane-bound physiological α-synuclein to highly aggregated forms. We then fractionated aqueous extracts (cytosol) from cerebral cortex using non-denaturing methods to search for soluble, disease-associated high molecular weight species potentially associated with toxicity. We applied these fractions and corresponding insoluble fractions containing Lewy-type aggregates to several reporter assays to determine their bioactivity and cytotoxicity. Ultimately, high molecular weight cytosolic fractions enhances phospholipid membrane permeability, while insoluble, Lewy-associated fractions induced morphological changes in the neurites of human stem cell-derived neurons. While the concentrations of soluble, high molecular weight α-synuclein were only slightly elevated in brains of dementia with Lewy bodies patients compared to healthy, age-matched controls, these observations suggest that a small subset of soluble α-synuclein aggregates in the brain may drive early pathogenic effects, while Lewy body-associated α-synuclein can drive neurotoxicity.
Sigma-Aldrich
Anti-α-Synuclein Antibody, clone SOY1, clone SOY1, from mouse
Sigma-Aldrich
N27 Rat Dopaminergic Neural Cell Line, The N27 rat dopaminergic neural cell line has been used as a dopaminergic neuron model for in vitro and in vivo studies.
Sigma-Aldrich
Anti-α-Synuclein Antibody, clone 2F12, clone 2F12, from mouse