Merck
  • PERK-mediated expression of peptidylglycine α-amidating monooxygenase supports angiogenesis in glioblastoma.
PERK-mediated expression of peptidylglycine α-amidating monooxygenase supports angiogenesis in glioblastoma.
Oncogenesis (2020-02-15)
Himanshu Soni, Julia Bode, Chi D L Nguyen, Laura Puccio, Michelle Neßling, Rosario M Piro, Jonas Bub, Emma Phillips, Robert Ahrends, Betty A Eipper, Björn Tews, Violaine Goidts
ABSTRACT
PKR-like kinase (PERK) plays a significant role in inducing angiogenesis in various cancer types including glioblastoma. By proteomics analysis of the conditioned medium from a glioblastoma cell line treated with a PERK inhibitor, we showed that peptidylglycine α-amidating monooxygenase (PAM) expression is regulated by PERK under hypoxic conditions. Moreover, PERK activation via CCT020312 (a PERK selective activator) increased the cleavage and thus the generation of PAM cleaved cytosolic domain (PAM sfCD) that acts as a signaling molecule from the cytoplasm to the nuclei. PERK was also found to interact with PAM, suggesting a possible involvement in the generation of PAM sfCD. Knockdown of PERK or PAM reduced the formation of tubes by HUVECs in vitro. Furthermore, in vivo data highlighted the importance of PAM in the growth of glioblastoma with reduction of PAM expression in engrafted tumor significantly increasing the survival in mice. In summary, our data revealed PAM as a potential target for antiangiogenic therapy in glioblastoma.
Sigma-Aldrich
RIPA Buffer
Pricing and availability is not currently available.
Sigma-Aldrich
EIF2AK3 Activator, CCT020312, The EIF2AK3 Activator, CCT020312 modulates the biological activity of EIF2AK3.
Pricing and availability is not currently available.