Merck
  • Dynamically crosslinked polymer nanocomposites to treat multidrug-resistant bacterial biofilms.
Dynamically crosslinked polymer nanocomposites to treat multidrug-resistant bacterial biofilms.
Nanoscale (2018-09-29)
Dong Yu Zhu, Ryan F Landis, Cheng-Hsuan Li, Akash Gupta, Li-Sheng Wang, Yingying Geng, Sanjana Gopalakrishnan, Jian Wei Guo, Vincent M Rotello
ABSTRACT
Multidrug-resistant biofilms are highly resistant to current antimicrobial therapies. We have developed an antimicrobial platform that integrates the bacteria-killing phytochemical carvacrol into dynamically crosslinked polymer nanocomposites (DCPNs). Taking advantage of a reversibly crosslinked Schiff-base scaffold throughout the engineered emulsions, DCPNs exhibited long-term shelf-life and good stability in serum, while readily disassembling in acidic microenvironments. Furthermore, we demonstrated that DCPNs efficiently penetrate the biofilm matrix, eradicating both Gram-negative/positive bacteria enclosed within. Moreover, DCPNs showed no observable toxicity to fibroblast mammalian cells with the same antimicrobial concentrations necessary to eradicate MDR biofilms. Given their potent antimicrobial and stimuli-responsive dissociation characteristics in a biofilm setting, DCPNs are a suitable therapeutic platform for combating MDR bacterial infections.
Sigma-Aldrich
Dichloromethyl methyl ether, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis
Pricing and availability is not currently available.