Merck
  • FANCA Promotes DNA Double-Strand Break Repair by Catalyzing Single-Strand Annealing and Strand Exchange.
FANCA Promotes DNA Double-Strand Break Repair by Catalyzing Single-Strand Annealing and Strand Exchange.
Molecular cell (2018-07-31)
Anaid Benitez, Wenjun Liu, Anna Palovcak, Guanying Wang, Jaewon Moon, Kevin An, Anna Kim, Kevin Zheng, Yu Zhang, Feng Bai, Alexander V Mazin, Xin-Hai Pei, Fenghua Yuan, Yanbin Zhang
ABSTRACT
FANCA is a component of the Fanconi anemia (FA) core complex that activates DNA interstrand crosslink repair by monoubiquitination of FANCD2. Here, we report that purified FANCA protein catalyzes bidirectional single-strand annealing (SA) and strand exchange (SE) at a level comparable to RAD52, while a disease-causing FANCA mutant, F1263Δ, is defective in both activities. FANCG, which directly interacts with FANCA, dramatically stimulates its SA and SE activities. Alternatively, FANCB, which does not directly interact with FANCA, does not stimulate this activity. Importantly, five other patient-derived FANCA mutants also exhibit deficient SA and SE, suggesting that the biochemical activities of FANCA are relevant to the etiology of FA. A cell-based DNA double-strand break (DSB) repair assay demonstrates that FANCA plays a direct role in the single-strand annealing sub-pathway (SSA) of DSB repair by catalyzing SA, and this role is independent of the canonical FA pathway and RAD52.
Sigma-Aldrich
MISSION® esiRNA, targeting human FANCA
Sigma-Aldrich
MISSION® esiRNA, targeting human FANCL
Sigma-Aldrich
Benzamidine hydrochloride, 99%
Sigma-Aldrich
MISSION® esiRNA, targeting human FANCB
Sigma-Aldrich
MISSION® esiRNA, targeting human FANCD2