
Vancomycin supplement

Synonym(s):
Vancomycin hydrochloride
Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · xHCl
CAS Number:
Molecular Weight:
1449.25 (free base basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing