
Valdecoxib

Synonym(s):
4-(5-Methyl-3-phenyl-4-isoxazolyl)benzenesulfonamide
Empirical Formula (Hill Notation):
C16H14N2O3S
CAS Number:
Molecular Weight:
314.36
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing