Skip to Content
Merck
TSKgel® G2500PW HPLC Column phase hydroxylated methacrylate, L × I.D. 30 cm × 21.5 mm, 17 μm particle size

TSKgel® G2500PW (17 µm) HPLC Columns

TSKgel® G2500PW HPLC Column phase hydroxylated methacrylate, L × I.D. 30 cm × 21.5 mm, 17 μm particle size
Synonym(s):
TSKgel® Size Exclusion (PW-Type) HPLC Column

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2