Skip to Content
Merck
Trimethoprim Related Compound B certified reference material, pharmaceutical secondary standard

Trimethoprim Related Compound B

Trimethoprim Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
(2,4-Diamino-5-pyrimidinyl)(3,4,5-trimethoxyphenyl)methanone, 5-(3,4,5-Trimethoxybenzoyl)-2,4-pyrimidinediamine
Empirical Formula (Hill Notation):
C14H16N4O4
CAS Number:
Molecular Weight:
304.30

Products

Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade