
Trifolirhizin

Empirical Formula (Hill Notation):
C22H22O10
CAS Number:
Molecular Weight:
446.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing