
Thiocolchicoside crystallised from ethanol

Synonym(s):
Thiocolchicoside, (S)-N-[3-(β-D-glucopyranosyloxy)-1,2-dimethoxy-10(methylthio)-9-oxo-5,6,7,9-tetrahydrobenzo [a] heptalen-7-yl] acetamide, 2-Demethoxy-2-glucosidoxythiocolchicine
Empirical Formula (Hill Notation):
C27H33NO10S
CAS Number:
Molecular Weight:
563.62
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing