
Sulfacetamide sodium

Synonym(s):
Sulfacetamide sodium salt monohydrate
Empirical Formula (Hill Notation):
C8H9N2NaO3S · H2O
CAS Number:
Molecular Weight:
254.24
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2