
Sucrose solution

Synonym(s):
OmniPur® Sucrose Solution, 20% W/V
Empirical Formula (Hill Notation):
C12H22O11
Molecular Weight:
342.30
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Format
Chemical Composition
Quality Segment
Technique
Showing 1-3 of 3