
(S)-(−)-Sulpiride

Synonym(s):
(S)-5-Aminosulfonyl-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxybenzamide, Levosulpiride
Empirical Formula (Hill Notation):
C15H23N3O4S
CAS Number:
Molecular Weight:
341.43
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing