Skip to Content
Merck
Reinecke salt

Reinecke salt

Reinecke salt
Synonym(s):
Ammonium reineckate, Ammonium tetrarhodanatodiamminechromate(III), Ammonium tetrathiocyanatodiamminechromate(III), Ammonium tetrathiocyanodiammonochromate
Linear Formula:
NH4[Cr(NH3)2(SCN)4]
CAS Number:
Molecular Weight:
336.43
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing