
(R)-BINAP

Synonym(s):
(R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene, (R)-(+)-(1,1′-Binaphthalene-2,2′-diyl)bis(diphenylphosphine), (R)-(+)-BINAP
Linear Formula:
[(C6H5)2PC10H6-]2
CAS Number:
Molecular Weight:
622.67
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing